Rubber Policeman
With glass rod
Catalog # 470104-462
Supplier: Avantor
Jump to:
Product Details & Documents
Ideal for Scraping and Mixing.
- Use as a policeman, stirring rod, or as a spatula
Rubber policeman is 15 mm natural rubber, fits 5 and 6 mm Ø rods. Glass rod is 15 cm long.
Specifications
- Description:With glass rod