Roman's Notes On DNA
Roman's Notes On DNA, 200, 20 cm, 25 cm
Catalog # 470101-028
Supplier: FITZHENRY & WHITESIDE LIMITED
Supplier #: 1895579937
Jump to:
Product Details & Documents
A consolidation of hints, and memory aids to help remember key concepts about DNA.
This book is an asset to any student or teacher searching for help in this complex topic.
Specifications
- Description:Roman's Notes On DNA
- Length:20 cm
- No. of pages:200
- Width:25 cm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



