Orders LinkContactUsLinkComponent
Reticular Tissue Slide
Reticular Tissue Slide
Reticular Tissue Slide, Section, Manual silver method and nuclear fast red
Catalog # 470182-632
Supplier:   Avantor
Reticular Tissue Slide
Catalog # 470182-632
Supplier:  Avantor
Reticular Tissue Slide
Reticular Tissue Slide
Reticular Tissue Slide, Section, Manual silver method and nuclear fast red
Price
$15.15
Log in for availability
Each
Product Details & Documents
Illustrates lymphatic system structures
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Stained to show reticular architecture of lymph node.
Specifications
  • Description:
    Reticular Tissue Slide
  • Stain:
    Manual silver method and nuclear fast red
  • Orientation:
    Section