Replacement Pump Oil
Replacement Oil for Vacuum Pumps, 946 mL
470104-444
: OIL-012
This is a specially prepared, low vapor pressure mineral oil. It is rust-inhibiting and foam resistant, and permits water condensates to drop out of suspension.
- Description:Replacement Oil for Vacuum Pumps, 946 mL