ContactUsLinkComponent
Ward's® Refractive Index Immersion Slide Set
Ward's® Refractive Index Immersion Slide Set
Catalog # 470024-872
Supplier:  Avantor
undefined
Ward's® Refractive Index Immersion Slide Set
Catalog # 470024-872
Supplier:  Avantor
Specifications
  • Description:
    Refractive Index Immersion Slide Set
  • No. of Slides:
    10
  • Cat. no.:
    470024-872
  • Supplier No.:
    WARD470024-872
Specifications
About this item
Ten New Slides in a Complete Teaching Set
  • Hand Made by Wards® staff Geologists
  • 10 Specially Selected Samples
  • Demonstrates Refractive Index of Minerals
Students learn to evaluate refractive index using the Becke Line method.
Set includes teacher’s guide and student worksheets.
10 slides with one mineral each: fluorite, microcline, albite, quartz, labradorite, biotite, calcite, hornblende, almandine, granite.