ContactUsLinkComponent
Reduce; Reuse; Recycle: A Plastics Identification Lab Activity
Reduce; Reuse; Recycle: A Plastics Identification Lab Activity
Catalog # 470004-352
Supplier:  Avantor
undefined
Reduce; Reuse; Recycle: A Plastics Identification Lab Activity
Catalog # 470004-352
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Reduce; Reuse; Recycle: A Plastics Identification Lab Activity
  • No. of Lab Groups:
    30
  • Cat. no.:
    470004-352
  • Supplier No.:
    WARD470004-352
Specifications
About this item
Make Your Students “Plastic Literate”
  • Learn to differentiate between different plastics
  • Linked to real world scenarios
  • Includes instructions
Increase students’ awareness of plastics in everyday use. Activities include interpreting recycling symbols on plastic materials, and performing scientific tests (density, alcohol, etc.) to identify various types of plastics. Includes materials for 30 students and instructions.