Red Tide Slide
Red Tide, Whole mount, Fast Green and Mallory's
Catalog # 470181-022
Supplier: Avantor
Jump to:
Product Details & Documents
Algal Bloom
- Identifying features clearly distinguishable
- Rigorous quality control standards
Wild bloom from the Northeast coast of the United States.
Specifications
- Description:Red Tide
- Stain:Fast Green and Mallory's
- Orientation:Whole mount