ContactUsLinkComponent
Force Probe Clamp
Force Probe Clamp
Catalog # 470134-172
Supplier:  Wards
undefined
Force Probe Clamp
Catalog # 470134-172
Supplier:  Wards
Supplier Number:  FCP-RED
Specifications
  • Description:
    Force Probe Clamp
  • Cat. no.:
    470134-172
  • Supplier No.:
    FCP-RED
Specifications
About this item
Provide a Secure Base for Force Probe Activities.
Attach any Force Probe to a force table, inclined plane, or table edge with this easy-to-use clamp accessory. The metal clamp opens from 13 to 39 mm, and attaches securely to surfaces of all sizes. The top rod is 30 mm tall and 9.8 mm in diameter.