Rectangular Supports
Supplier: Shiv Dial Sud & Sons
Jump to:
About this item
Tip-free stability for experiments.
- Glassware can sit directly on base
- Rods are zinc-plated steel
Heavy cast iron bases with black enamel finish. Includes a removable zinc-plated steel rod.
2 Options Available Below
Product Details & Documents
Tip-free stability for experiments.
- Glassware can sit directly on base
- Rods are zinc-plated steel
Heavy cast iron bases with black enamel finish. Includes a removable zinc-plated steel rod.
Product Family Options
Product Information
- Base dimensionsRod diameterRod lengthAvailabilityPrice
- Supplier Number: SSB4X6-CI
- 5×8"⅜"20"Each$39.60Case of 6$237.60Supplier Number: CH12321/2