If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:WARD’S Qualitative Water Analysis Lab Activity
- Cat. no.:470175-710
- Supplier No.:WARD470175-710
Test Collected Samples for Common Industrial and Biological Contaminants
Students can detect nitrates, iron, phosphates, chloride, ammonia, pH, hard water, sulfate, and chlorine in water samples, and compare samples from a variety of environments. Each test is performed by a separate lab station. It comes with enough materials for ten setups, a teacher’s guide, and student copymasters.
Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.