Orders LinkContactUsLinkComponent
PYREX® Student Burets with Replaceable Components
Supplier:   Corning
PYREX® Student Burets with Replaceable Components
Supplier:  Corning
Certificates
PYREX® Student Burets with Replaceable Components
PYREX® Student Burets with Replaceable Components
2 Options Available Below
Product Details & Documents
Durable Enough for Repeated Student Use.
  • Tooled tops and bottoms
  • White enamel graduations
  • Teflon® stopcock reduces breakage
The replaceable stopcock assemblies feature a screw thread locking nut and collar and a replaceable tip with ground end. Teflon® stopcock permits replacement of the body or tip. New triangular shipping package reduces breakage in transit. Replacement parts available.
Product Family Options
Product Information
  • Capacity
    Graduation Range
    Availability
    Price
  • 470211-384
    Pyrex® Replaceable Buret
    Pyrex® Replaceable Buret
    25 ml
    0.10 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $400.00
    $400.00
    Log in for availability
    Case of 6
    Case of 6 - $1,200.00
    $1,200.00
    Pack of 2
       $400.00
    Case of 6
       $1,200.00
    Supplier Number: 2116-25
  • 470211-392
    Pyrex® Replaceable Buret Tip
    Pyrex® Replaceable Buret Tip
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $16.20
    $16.20
    Log in for availability
    Case of 12
    Case of 12 - $194.40
    $194.40
    Each
       $16.20
    Case of 12
       $194.40
    Supplier Number: 2116-TO