Orders LinkContactUsLinkComponent
PYREX® Serological Pipettes, Plugged
PYREX® Serological Pipettes, Plugged
Supplier:   Corning
PYREX® Serological Pipettes, Plugged
Supplier:  Corning
Certificates
PYREX® Serological Pipettes, Plugged
PYREX® Serological Pipettes, Plugged
3 Options Available Below
Product Details & Documents
These sterile, glass pipettes, calibrated to deliver with blow-out; offer long slender tapered tips to make pipetting faster and easier.
Color-coded sizes.
Capacity Negative Grad. Divisions
WLS-69650-C 1.0 ml 0.2 ml 0.01 ml
WLS-69650-G 5.0 ml 1.0 ml 0.1 ml
WLS-69650-H 10.0 ml 2.0 ml 0.01 ml.
Documents
Product Family Options
Product Information
  • Capacity
    Negative graduations
    Subdivisions
    Packed
    Availability
    Price
  • 470211-592
    Pipet, sterile serological Pyrex®
    Pipet, sterile serological Pyrex®
    2 ml
    0.2 ml
    0.01 ml
    Individually Packed
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 180
    Pack of 180 - $272.00
    $272.00
    Log in for availability
    Case of 720
    Case of 720 - $1,088.00
    $1,088.00
    Pack of 180
       $272.00
    Case of 720
       $1,088.00
    Supplier Number: 7077-2N
  • 470211-594
    Pipet, sterile serological Pyrex®
    Pipet, sterile serological Pyrex®
    5 ml
    1.0 ml
    0.1 ml
    Individually Packed
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 120
    Pack of 120 - $213.00
    $213.00
    Log in for availability
    Case of 720
    Case of 720 - $1,278.00
    $1,278.00
    Pack of 120
       $213.00
    Case of 720
       $1,278.00
    Supplier Number: 7077-5N
  • 470211-596
    Pipet, sterile serological Pyrex®
    Pipet, sterile serological Pyrex®
    5 ml
    1.0 ml
    0.1 ml
    Bulk Packed
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $46.00
    $46.00
    Log in for availability
    Case of 960
    Case of 960 - $1,472.00
    $1,472.00
    Pack of 30
       $46.00
    Case of 960
       $1,472.00
    Supplier Number: 7078-5N