PYREX® Precision Burets
: Corning
Suitable for Use Laboratories Not Requiring the Accuracy of Class A Models.
- Dispensing Burets
- Permanent graduation markings
- Nonstick 2 mm Teflon stopcock ensures a smooth, leak-free operation
- Dust cover included
Product Family Options
- CapacityGraduation Interval
- : 2122A-25
- : 2122A-50