PYREX® Knob Top Bell Jar
Catalog # 470211-570
Supplier: Corning
Specifications
- Description:Bell Jar, Pyrex, w/ Knob Top, 16.5 cm x 28 cm, 4 L Capacity, Corning
- Environmentally Preferable:
- Cat. no.:470211-570
- Supplier No.:6885-165
Specifications
About this item
These specially mold blown jars have a fine ground finish bottom flange to assure tight sealing with ground glass plates.
Caution: Do not use with heat, pressure or vacuum applications.