PYREX® Filtering Flasks
Supplier: Corning
About this item
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks have a beaded top that provides an excellent fit for rubber stoppers. Graduated to show approximate volume.
The heavy-duty rim flange is ideal for supporting Buchner funnels. It includes a side arm for attaching rubber tubing. Extra-large marking spot for easy identifications.
3 Options Available Below
Product Details & Documents
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks have a beaded top that provides an excellent fit for rubber stoppers. Graduated to show approximate volume.
The heavy-duty rim flange is ideal for supporting Buchner funnels. It includes a side arm for attaching rubber tubing. Extra-large marking spot for easy identifications.
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
- Catalog No:
- 470211-504
- 470211-506
- 470211-502
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
- CapacityAvailabilityPrice
- 250 mlPack of 6$271.00Case of 18$813.00Supplier Number: 5340-250Specifications:
More
Specifications
Cat. No.470211-504Environmentally PreferableEnvironmental certification - 500 mlPack of 6$379.00Case of 18$1,137.00Supplier Number: 5340-500Specifications:
More
Specifications
Cat. No.470211-506Environmentally PreferableEnvironmental certification - 1000 mlEach$118.00Case of 12$1,416.00Supplier Number: 5340-1LSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.470211-502Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - MonterreyLow Manufacturing Impact