Orders LinkContactUsLinkComponent
PYREX® Burets
   Sustainable Options Available
Supplier:   Corning
PYREX® Burets
Supplier:  Corning
   Sustainable Options Available
Certificates
PYREX® Burets
3 Options Available Below
Product Details & Documents
Educational grade Pyrex burets.
  • Heavy duty
  • Transparent
Supplied with self-lubricating, leakproof PMP stopcock. This buret is unaffected by most dilute aqueous solutions, except ammonia and HF. It is not recommended for use with ethanol or organic solvents. The clear white markings make dark liquids easier to measure the the Teflon stopcock is less likely to freeze up.
Specifications for Product Family
Specifications
  • Catalog No:
  • 470211-386
  • 470211-390
  • 470146-252
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
  • Capacity
    Graduation Interval
    Availability
    Price
  • Geissler Buret
    50 ml
    0.1 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - 209,00 $
    209,00 $
    Log in for availability
    Case of 6
    Case of 6 - 1 254,00 $
    1 254,00 $
    Each
       209,00 $
    Case of 6
       1 254,00 $
    Supplier Number: 2116-50
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470211-386
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 470211-390
    Geissler Buret Locking Stopcock
    Geissler Buret Locking Stopcock
    Log in for availability
    Log in for availability
    Each
    Each - 163,00 $
    163,00 $
    Each
       163,00 $
    Supplier Number: 2116-LSO
    Specifications:
    More
    Specifications
    Cat. No.
    470211-390
    Environmentally Preferable
    Environmental certification
  • 470146-252
    Geissler buret replacement tip, pack of 12, six grinded tips and six clear (ungrinded) tipss
    Geissler buret replacement tip, pack of 12, six grinded tips and six clear (ungrinded) tipss
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - 197,00 $
    197,00 $
    Pack of 12
       197,00 $
    Supplier Number: 2116-GTO2
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    470146-252
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact