Order Entry
Canada
ContactUsLinkComponent
Pulleys in Motion Kit
Pulleys in Motion Kit
Catalog # 470101-914
Supplier:  SI Manufacturing
undefined
Pulleys in Motion Kit
Catalog # 470101-914
Supplier:  SI Manufacturing
Supplier Number:  37480

Specifications

  • Description:
    Kit, Pulleys in Motion
  • Cat. no.:
    470101-914
  • Supplier No.:
    37480

Specifications

About this item

Versatile teaching kit contains enough materials for the whole classroom to learn about and experiment with a variety of pulley configurations. Because these activities are hands-on, students get a real feel for the forces at work.

For example, they will feel the difference between a single and a multiple pulley system when they use the two systems to lift the same load. Covers concepts of work, power, mass and more.

Delivery: Includes 18 single-sheave pulleys, 12 double-sheave pulleys, 12 triple-sheave pulleys, six spring scales (500 g/5 N), roll of twine (107 m/350 ft.), 20 pulley chain links, and Pulleys In Motion Teacher’s Guide.
Caution: WARNING: CHOKING HAZARD- small parts not for children under 3 years.