Protractor
Protractor, Plastic, 180° 10 cm; 6" Ruler
Catalog # 470002-858
Supplier: Skywill Far East Industries
Supplier #: 5010
Jump to:
Product Details & Documents
Six inch clear plastic protractor for measuring 180° in 1° divisions for both left and right opening angles.
The top and bottom of the straight edge are graduated in both cm and inches respectively.
Specifications
- Description:Protractor, Plastic, 180° 10 cm; 6" Ruler
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



