Order Entry
Canada
Orders LinkContactUsLinkComponent
Protractor

Protractor

Protractor, 4"
Catalog # 470235-440
Supplier:   Acme United
Supplier #:  259
Protractor
Catalog # 470235-440
Supplier:  Acme United
Protractor
Protractor

Protractor, 4"

Price
$51.27
Log in for availability
Box of 12 Case of 144
Product Details & Documents

Made of laminated, transparent plastic with solid center. Graduated 0 to 360° and 0 to 90° to 0 in 1° intervals.

Specifications
  • Description:
    Protractor
  • Diameter:
    4"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.