ContactUsLinkComponent
Proteus vulgaris Slide
Proteus vulgaris Slide
Catalog # 470180-866
Supplier:  Avantor
undefined
Proteus vulgaris Slide
Catalog # 470180-866
Supplier:  Avantor
Specifications
  • Description:
    Proteus vulgaris
  • Cat. no.:
    470180-866
  • Stain:
    Gram (-)
  • Supplier No.:
    WARD470180-866
  • Orientation:
    Smear
Specifications
About this item
Species of Proteobacteria
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Proteus hauseri. Rods. It causes urinary tract infections in humans.