Orders LinkContactUsLinkComponent
Protein InstaStain®
Protein InstaStain®
Protein InstaStain®, For 15 Gels
Catalog # 470002-532
Supplier:   Edvotek
Supplier #:  2016
Protein InstaStain®
Catalog # 470002-532
Supplier:  Edvotek
CAS Number:  
Protein InstaStain®
Protein InstaStain®
Protein InstaStain®, For 15 Gels
CAS Number:  
Price
$133.85
Log in for availability
Each
Product Details & Documents
High Quality Results, Excellent for Photodocumentation
Stain protein gels in just 20 minutes, plus fixing and destaining times. Protein InstaStain® is far faster than liquid blue stains and ensures a yield of uniform gel results. Environmentally friendly InstaStain® also uses a solid matrix to avoid large amounts of liquid waste. For 15 gels.
Specifications
Specifications
  • Description:
    Protein InstaStain®, For 15 Gels