ContactUsLinkComponent
Project STAR Spectrometer
Project STAR Spectrometer
Catalog # 470003-848
Supplier:  Science First
undefined
Project STAR Spectrometer
Catalog # 470003-848
Supplier:  Science First
Supplier Number:  614-0210
Specifications
  • Description:
    Project STAR Spectrometer
  • Cat. no.:
    470003-848
  • Supplier No.:
    PS14-PLAS
Specifications
About this item
View Spectra with an Accuracy of ±1 Nanometer
Examine the spectra of the Sun and stars, and those produced by minerals in flame tests. It features a high-quality grating and stepped slit for a clear, precise, spectrum. The sturdy plastic construction is labeled with both electron volts and nanometers, as well as a chart of common spectral lines. An instruction manual is included. Size: 12"L x 8 1/2"W.