ContactUsLinkComponent
Direct Vision Spectroscope, Advanced
Direct Vision Spectroscope, Advanced
Catalog # 470123-418
Supplier:  Wards
undefined
Direct Vision Spectroscope, Advanced
Catalog # 470123-418
Supplier:  Wards
Supplier Number:  PH0594A
Specifications
  • Description:
    Direct vision spectroscope, advanced
  • Size:
    25 Ø×96 mm L
  • Cat. no.:
    470123-418
  • Supplier No.:
    PH0594
Specifications
About this item
Fine hand instrument for qualitative visible spectrum analysis.
  • Features adjustable slit width and draw-out focusing collar
  • Uses 3-element prisms in lieu of diffraction grating
  • Comes in an attractive and robust wooden case
Handy spectroscope for viewing the visible spectra of light sources. Wavelength dispersion is accomplished by a fixed prism rather than a diffraction grating, so the spectrum may be viewed directly rather than at an oblique angle. With an adjustable collar, the metal tube may be drawn out to focus. An adjustable light slit is fitted with achromatic objective glass and 3 element prisms. Comes complete in a wooden case for safe storage.