Precision Test Strips
Precision Test Strips, Test: Nitrate, Range, ppm: 0 - 250, Color Chart Concentrations: 0, 10, 25, 50, 100, 250, 500, 0 - 250 ppm
Supplier: Precision Laboratories
Supplier #: NAT-1V-50
Jump to:
Product Details & Documents
Precision Test Strips quickly test for low levels of nitrite and chlorine.
- Fast Tests
- Pool Test Strip Measures pH, free, and Total Chlorine
- Aquarium Test Strip Measures pH and Chlorine Levels
- Fifty Test Strips in One Package
- Test Strips for Nitrite and Chlorine
After immersing the strip in the solution to be measured, the concentration is measured by comparing the strip’s color to a color chart on the package.
The Pool Test Strip is three tests in one—pH, free, and total chlorine. The aquarium test strip simultaneously measures pH and chlorine levels.
Packaging: Each package includes 50 test strips in a resealable package.
Documents
Specifications
Specifications
- Description:Precision Test Strips, Test: Nitrate, Range, ppm: 0 - 250, Color Chart Concentrations: 0, 10, 25, 50, 100, 250, 500
- Range:0 - 250 ppm