Order Entry
Canada
Orders LinkContactUsLinkComponent
Potassium Hydroxide

Potassium Hydroxide

Supplier:   Ward's Science
Potassium Hydroxide
Supplier:  Ward's Science
Certificates
Potassium Hydroxide
Potassium Hydroxide

12 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 1310-58-3
Formula Weight: 56.11
Formula: KOH
Hazard Info: Corrosive, Irritant, Toxic
Density (g/mL): 2.044
Boiling Point (°C): 1320
Freezing Point (°C): 361
Solubility: Water, Alcohol, and Glycerin
Synonyms: Caustic Potash, Potassium Hydrate
Shelf Life (months): 36
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Product Family Options

Product Information

  • Grade
    Size
    Chemical/physical specifications
    Availability
    Price
  • 470302-142
    Potassium Hydroxide
    Potassium Hydroxide
    Reagent
    100 g
    Pellet
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $22.45
    $22.45
    Each (100g)
       $22.45
    Minimum order quantity is 1

    Supplier Number: PP0570-100GR
  • 470302-144
    Potassium Hydroxide
    Potassium Hydroxide
    Reagent
    500 g
    Pellet
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $66.75
    $66.75
    Each (500g)
       $66.75
    Minimum order quantity is 1

    Supplier Number: PP0570-500GR
  • 470302-134
    Potassium Hydroxide
    Potassium Hydroxide
    Laboratory
    500 g
    Flake
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $52.00
    $52.00
    Each (500g)
       $52.00
    Minimum order quantity is 1

    Supplier Number: PP0590-500G
  • 470302-132
    Potassium Hydroxide
    Potassium Hydroxide
    Laboratory
    2.5 kg
    Flake
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $277.90
    $277.90
    Each (2,500g)
       $277.90
    Minimum order quantity is 1

    Supplier Number: PP0590-2.5KG
  • 470045-602
    Potassium Hydroxide
    Potassium Hydroxide
    Laboratory
    100 g
    Pellet
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $23.05
    $23.05
    Each (100g)
       $23.05
    Minimum order quantity is 1

    Supplier Number: PP0570-100G
  • 470302-140
    Potassium Hydroxide
    Potassium Hydroxide
    Laboratory
    500 g
    Pellet
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $59.00
    $59.00
    Each (500g)
       $59.00
    Minimum order quantity is 1

    Supplier Number: PP0570-500G
  • 470302-138
    Potassium Hydroxide
    Potassium Hydroxide
    Laboratory
    2.5 kg
    Pellet
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $259.05
    $259.05
    Each (2,500g)
       $259.05
    Minimum order quantity is 1

    Supplier Number: PP0570-2.5KG
  • 470233-554
    Potassium Hydroxide, Solution
    Potassium Hydroxide, Solution
    Laboratory
    30 mL
    15%
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $21.80
    $21.80
    Pack of 5
       $21.80

    Supplier Number: PP0599-PK5
  • 470225-680
    Potassium Hydroxide, Solution
    Potassium Hydroxide, Solution
    Laboratory
    500 mL
    0.1 M
    Log in for availability
    Log in for availability
    Each
    Each - $11.60
    $11.60
    Each
       $11.60

    Supplier Number: PP0597-500ML
  • 470302-146
    Potassium Hydroxide, Solution
    Potassium Hydroxide, Solution
    Laboratory
    1 L
    0.1 M
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $14.45
    $14.45
    Each (1l)
       $14.45
    Minimum order quantity is 1

    Supplier Number: PP0600-1L
  • 470302-148
    Potassium Hydroxide, Solution
    Potassium Hydroxide, Solution
    Laboratory
    1 L
    1 M
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $32.00
    $32.00
    Each (1l)
       $32.00
    Minimum order quantity is 1

    Supplier Number: PP0591-1L
  • 470302-130
    Potassium Hydroxide, QuickPrep
    Potassium Hydroxide, QuickPrep
    Laboratory
    Makes 500 mL
    1 M
    Log in for availability
    Log in for availability
    Each (50ml)
    Each (50ml) - $19.50
    $19.50
    Each (50ml)
       $19.50
    Minimum order quantity is 1

    Supplier Number: PP0570-CONC

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.