Potassium Acetate
Potassium Acetate, Reagent, 500 g
Supplier: Ward's Science
: PP0330-500GR
Jump to:
Product Details & Documents
CAS Number: 127-08-2
Formula Weight: 98.14
Formula: CH3COOK
Density (g/mL): 1.57-1.8
Freezing Point (°C): 292
Solubility: Water
Synonyms: Acetic Acid Potassium Salt
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Specifications
Specifications
- Description:Potassium Acetate
- Size:500 g
- Chemical/physical specifications:Crystal
- Grade:Reagent



