ContactUsLinkComponent
Polypropylene Petri Dish Rack
Polypropylene Petri Dish Rack
  470020-886
 :  Bel-Art
undefined
Polypropylene Petri Dish Rack
  470020-886
 :  Bel-Art
 :  F18991-0000_SE
  • Description:
    Polypropylene Petri Dish Rack
  • Cat. no.:
    470020-886
  • Supplier No.:
    F189910000-B
This autoclavable polypropylene rack holds forty-two 100 mm dishes. It has a small footprint, is stackable to conserve counter space, and it features a handle for easy transport. Size: 13"L x 8 1/4"W x 7"H.