Polypropylene Beaker Set
Catalog # 490012-164
Supplier: Dynalon
Specifications
- Capacity:50 ml, 100 ml, 250 ml, 500 ml, 1000 ml
- Description:Set of 5
- Cat. no.:490012-164
- Supplier No.:222195
Specifications
About this item
Unbreakable polypropylene.
- Autoclavable
- Chemical-resistant
These beaker sets autoclavable up to 145 °C and chemical resistant. All feature pour spouts and metric graduation scales.