Polypropylene Beaker Set
50 ml, 100 ml, 250 ml, 500 ml, 1000 ml, Set of 5
470122-772
: BPSET5-SW
Made of unbreakable polypropylene, this beaker set avoids dangerous accidents.
- Beakers Can Withstand up to 120°C
- Autoclavable
- Permanent Graduations
- Unbreakable
- No-Drip Pouring Spouts
The beakers are ideal for educational use and feature permanent graduations and no-drip pouring spouts.
They are autoclavable and can withstand temperatures up to 120°C.
: These beakers come in a polyethylene bag.
- Capacity:50 ml, 100 ml, 250 ml, 500 ml, 1000 ml
- Description:Set of 5
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



