Orders LinkContactUsLinkComponent
PolyPaper Labels
PolyPaper Labels
PolyPaper Labels
PolyPaper Labels
Pressure-sensitive coated polyethylene labels are chemical, mildew, age, tear, and water resistant.
  • Polyolefin labels will not wrinkle, curl or melt
  • Waterproof and compatible with most lab chemicals
They accept a variety of inks including copier, marker, ballpoint, gels and technical markers.
Product Family Options
  • Description
    Material
    Dimensions
    Packaging
  • Polyolefin labels
    Polyolefin
    20.3×38.1 mm (0.8×1.5")
    64 labels/sheet
     : 6319-0015
  • Polyolefin labels
    Polyolefin
    22.9×22.9 mm (0.9×0.9")
    80 labels/sheet
    Pack of 800
    Pack of 800 - 254,00 $
    254,00 $
    Case of 4000
    Case of 4000 - 1 270,00 $
    1 270,00 $
    Pack of 800
       254,00 $
    Case of 4000
       1 270,00 $
     : 6319-0010