Polygrid Petri Dish Carrying Rack
Catalog #: 470105-610
Supplier: Bel-Art
Specifications
- Description:Petri Dish Carrying Rack, Poxygrid Holds 20 Dishes
- Cat. no.:470105-610
- Supplier No.:H189780000
Specifications
About this item
This epoxy coated steel wire rack holds 20 plastic petri dishes. Petri dishes are easily loaded and emptied from the top. The wide front opening permits easy access but prevents dishes from sliding out. Not suitable for glass petri dishes.
Customers Who Bought This Also Bought