Orders LinkContactUsLinkComponent
Plastic Dropping Pipettes
Plastic Dropping Pipettes
Plastic dropping pipettes
Catalog # 470146-305
Supplier:   GSC International
Supplier #:  1300-12A-DZ
Plastic Dropping Pipettes
Catalog # 470146-305
Supplier:  GSC International
Plastic Dropping Pipettes
Plastic Dropping Pipettes
Plastic dropping pipettes
Price
$10.90
Log in for availability
Pack of 12 Case of 144
Product Details & Documents
Drawn to a small outlet and fire-polished, these plastic pipettes with bulb are 12.5 cm in length.
Packaging: Package of 12.
Specifications
  • Description:
    Plastic dropping pipettes