Plastic Droppers
Plastic Dropper
Catalog # 470145-614
Supplier: Skywill Far East Industries
Supplier #: 2921400
Jump to:
Product Details & Documents
Perfect for student use.
- Each pack contains six medicine droppers with rubber bulbs
The total length of the dropper is 99 mm and capacity of each press is approximately 0.5 ml maximum.
Caution: WARNING: CHOKING HAZARD- Small Parts. Not for Children Under 3 Years.
Specifications
- Description:Plastic Dropper
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



