ContactUsLinkComponent
Ward's® Phenolphthalein Blood Test Kit
Ward's® Phenolphthalein Blood Test Kit
Catalog # 470024-156
Supplier:  Ward's Science
undefined
Ward's® Phenolphthalein Blood Test Kit
Catalog # 470024-156
Supplier:  Ward's Science
Supplier Number:  DD6001-KIT
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Phenolphthalein Presumptive Blood Test Kit
  • No. of Lab Groups:
    30
  • Cat. no.:
    470024-156
  • Supplier No.:
    DD6001-KIT
Specifications
About this item
Includes a known blood standard to monitor results.
  • Grades 9-12
  • Estimated class/lab time required is 30 minutes
Save time in laboratory experiments using the premixed chemicals and plastic dropper bottles of liquid reagents in this kit. Everything you need for the presumptive determination of real animal blood is included in a plastic storage case, with enough materials to conduct 30 tests. Note: The phenolphthalein test will not distinguish between human and animal blood. Further serology analysis is necessary.