Orders LinkContactUsLinkComponent
Peat Moss
Peat Moss
Peat Moss
Catalog # 470153-626
Supplier:   ARETT SALES CORP
Supplier #:  H28-15503
Peat Moss
Catalog # 470153-626
Supplier:  ARETT SALES CORP
Peat Moss
Peat Moss
Peat Moss
Price
$34.65
Log in for availability
Pack of 1 Case of 6
Product Details & Documents
Mixed in soil, it improves moisture retention and aeration, as well as increases acidity. Use as a starter for seeds, rootings, transplants, and cuttings.
Quantity: 11 liters.
Specifications
  • Description:
    Peat Moss