PCR Rack with Lid
Catalog # 470200-670
Supplier: Ward's Science
Specifications
- Description:PCR tube rack
- Cat. no.:470200-670
- Supplier No.:241000
Specifications
About this item
Organize and Store PCR Products or Reagents
This sturdy plastic rack features a 96 x 0.2 ml well format with an 8 x 12 grid, labeled for easy identification and record keeping. A lid securely locks tubes includes to prevent accidental spilling. Rack can be frozen to -20°C. Designed for use with most 0.2ml strip tubes or individual tubes.