ContactUsLinkComponent
Polymerase Chain Reaction (PCR) Simulation Kit
Polymerase Chain Reaction (PCR) Simulation Kit
  470039-554
 :  Edvotek
undefined
Polymerase Chain Reaction (PCR) Simulation Kit
  470039-554
 :  Edvotek
 :  103
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    PCR- Polymerase Chain Reaction Simulation Kit
  • Cat. no.:
    470039-554
  • Supplier No.:
    103
Ideal for Beginning PCR Instruction
This easy-to-use simulation activity introduces students to the indispensable techniques, principles, and applications of the Polymerase Chain Reaction (PCR) without the need for a thermal cycler. Students will develop the skills needed for further application and study of PCR. Ready-to-Load™ DNA samples are included to allow for quick and easy setup. Kit also includes UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, InstaStain® Blue and FlashBlue™ stain, calibrated pipette, 100 ml graduated cylinder, and microtipped transfer pipettes. Activity requires an electrophoresis apparatus, power supply, automatic micropipette and tips, balance, microwave or hot plate, and visualization system. Material supplied for six gels. Time requirements: 60 minutes.