Orders LinkContactUsLinkComponent
Osteogenesis, Endochondral Ossification Slide
Osteogenesis, Endochondral Ossification Slide
Osteogenesis, Endochondral Ossification Slide, Longitudinal section, Mallory Triple
Catalog # 470176-956
Supplier:   Avantor
Osteogenesis, Endochondral Ossification Slide
Catalog # 470176-956
Supplier:  Avantor
Osteogenesis, Endochondral Ossification Slide
Osteogenesis, Endochondral Ossification Slide
Osteogenesis, Endochondral Ossification Slide, Longitudinal section, Mallory Triple
Price
$15.65
Log in for availability
Each
Product Details & Documents
Fetal skeletal development
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal long bone. Extensive deposition of bony spicules with both per- and endochondral osteoblasts and osteoclasts.
Specifications
  • Description:
    Osteogenesis, Endochondral Ossification Slide
  • Stain:
    Mallory Triple
  • Orientation:
    Longitudinal section