Explorer® Precision Balances, Ohaus®
Catalog # 470203-742
Supplier: OHAUS CORP.
Specifications
- Model:EX2202
- Pan/Platform Size:190×200 mm
- Cat. no.:470203-742
- Readability:0.01 g
- Weighing Capacity:2200 g
- Supplier No.:EX2202
Specifications
About this item
Explorer precision balances offer intelligent performance.
- Fast Stabilization Times and Optimized Linearity
- Repeatability Specifications
- Sturdy, Modular Design
- Large Color Touchscreen
- Two-Year Manufacturer's Warranty
An innovative modular design features a color touch display that can be separated from the weighing base for convenient control over all modes of operation. SmarText™ 2.0 graphical software powers an intuitive user interface featuring 14 applications, virtual QWERTY and numeric keypads, and below-minimum weight applications.
The included draft shield provides ample access and visibility to the weighing chamber and features antistatic coated glass (only available with EX2202 model).
Ordering information: Two-year manufacturer's warranty is included with purchase.
Caution: Caution:Choking Hazard - Small parts. Not for children under 3 years.