Order Entry
Canada
Orders LinkContactUsLinkComponent
Noyes Iris Scissors

Noyes Iris Scissors

Noyes Iris Scissors, 110 mm (4.5"), Stainless steel, Sharp/Sharp
Catalog # 470018-936
Supplier:   Dunrite Instruments
Supplier #:  140976
Noyes Iris Scissors
Catalog # 470018-936
Noyes Iris Scissors

Noyes Iris Scissors, 110 mm (4.5"), Stainless steel, Sharp/Sharp

Price
$47.00
Log in for availability
Each
Product Details & Documents

Aligning guide point, fingertip control, and extremely fine blades aid in precise dissecting such as trimming tissue from nerves.

Specifications
  • Description:
    Noyes Iris Scissors
  • Length:
    110 mm (4.5")
  • Material:
    Stainless steel
  • Blade type:
    Sharp/Sharp
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.