ContactUsLinkComponent
Non-Scratching Duster for Lab Equipment
  470006-808
 :  BRUSHTECH
undefined
Non-Scratching Duster for Lab Equipment
  470006-808
 :  BRUSHTECH
 :  B265C
  • Bristle Color:
    Yellow
  • Bristle Material:
    Synthetic
  • Handle Material:
    Plastic
  • Description:
    Brush Duster, Non-Scratching
  • Cat. no.:
    470006-808
  • Handle Length:
    12 cm
  • Handle Color:
    White
  • Supplier No.:
    B265C
  • Brush Head Length:
    4 cm
Non-scratching duster for microscopes, laboratory equipment, printed circuits and other delicate equipment and instruments.