Medullated Nerve, Cross/Longitudinal Section, Masson Stained Slides
Catalogue # 470182-696
Fournisseur: Avantor
Spécifications
- Description:Nervous Tissue, Medullated Nerve
- Cat. no.:470182-696
- Stain:Mason
- Supplier No.:WARD470182-696
- Orientation:Cross section & longitudinal section
Spécifications
À propos de cet article
Demonstrates a Variety of Nervous Tissue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Peripheral nerve bundles showing axis cylinders, medullary sheaths, Schwann nuclei, epi- and perineurium.