Order Entry
Canada
ContactUsLinkComponent
The Nervous System Poster
The Nervous System Poster
Catalog # 470112-292
undefined
The Nervous System Poster
Catalog # 470112-292
Supplier Number:  1587790440

Specifications

  • Width:
    50 cm
  • Description:
    Nervous System Poster
  • Length:
    66 cm
  • Cat. no.:
    470112-292
  • Finish:
    Laminated
  • Supplier No.:
    1587790440

Specifications

About this item

Frontal view with central nervous structures in yellow and autonomic structures in white.

  • Five additional insets included