TeacherGeek Multi-Cutter - Easy Engineering
Catalog # 470132-516
Supplier: TEACHERGEEK INC
Specifications
- Description:TeacherGeek multi-cutter
- Cat. no.:470132-516
- Supplier No.:60000
Specifications
About this item
The Easy Cutter is designed to cut a variety of materials and components such as wood strips, connector strips, dowels and more.
- Design accommodates large and small hands, with a vinyl grip for comfortable handling
- Cutting plane includes marked angles for precision
The die-cast aluminum angle plate insures accurate angle cuts. These cutters are an essential tool in the TeacherGeek building system.
Adult supervision required - sharp edge.