Mold Types, Composite Slide
Mold Types, Composite Slide, Whole mount, Fast Green and Mallory's
Catalog # 470177-438
Supplier: Avantor
Jump to:
Product Details & Documents
Observe the differences between common mold types
- Identifying features clearly distinguishable
- Rigorous quality control standards
Aspergillus, Penicillium, and Rhizopus on the same slide, clearly contrasting sporangia and conidia.
Specifications
- Description:Mold Types, Composite Slide
- Stain:Fast Green and Mallory's
- Orientation:Whole mount