ContactUsLinkComponent
Mohr Buret Pinchcock Attachment
Mohr Buret Pinchcock Attachment
Catalog # 470161-250
Supplier:  Avantor
undefined
Mohr Buret Pinchcock Attachment
Catalog # 470161-250
Supplier:  Avantor
Specifications
  • Description:
    Pinchcock Attachment for Mohr Burets, for 60800 Series
  • Cat. no.:
    470161-250
  • Supplier No.:
    WARD470161-250
Specifications
About this item
Designed for the Flint Glass Mohr Burets. Consists of a short piece of rubber tubing, glass buret tip, and a pinchcock clamp. Easily slips over the end of any buret not having a stopcock.