Mirrors
Supplier: United Scientific Supplies
Jump to:
About this item
Traditional glass mirrors.
- Plate glass
- Silvered back
Use these top quality silvered mirrors for demonstrations and experiments in optics. Concave and convex mirrors are 50 mm dia.
2 Options Available Below
Product Details & Documents
Traditional glass mirrors.
- Plate glass
- Silvered back
Use these top quality silvered mirrors for demonstrations and experiments in optics. Concave and convex mirrors are 50 mm dia.
Product Family Options
Product Information
- AvailabilityPrice
- 470020-572Glass mirror, 4x4"Glass mirror, 4x4"Each$7.80Pack of 12$71.00Supplier Number: MSG4X4
- 470325-672Mirror, Plane, Glass, 6×2"Supplier Number: MSG6X2