ContactUsLinkComponent
Mini-Prep Isolation of Plasmid DNA
Mini-Prep Isolation of Plasmid DNA
Catalog # 470039-970
Supplier:  Edvotek
undefined
Mini-Prep Isolation of Plasmid DNA
Catalog # 470039-970
Supplier:  Edvotek
Supplier Number:  202-A
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Mini-Prep Isolation of Plasmid DNA
  • Environmentally Preferable:
  • Cat. no.:
    470039-970
  • Supplier No.:
    202-A
Specifications
About this item
Allow students to practice small-scale rapid isolation of plasmid DNA, which is a routine procedure used for screening and analysis of recombinant DNA in cloning and subcloning experiments.
In this experiment, students isolate plasmid DNA in just 60 minutes, without the use of toxic chemicals such as phenol or chloroform. Material provided can support up to 40 isolations and five DNA agarose gels. Kit includes instructions, Plasmid Extraction LyphoCells, gel loading solution, UltraSpec-Agarose powder, electrophoresis buffer, InstaStain FlashBlue and FlashBlue Plus stain, calibrated pipet, 100 ml graduated cylinder and microtipped transfer pipettes.
Ordering information: An electrophoresis tank, power supply, variable micropipet and tips, balance and microwave or hot plate are required and available seperately.