Orders LinkContactUsLinkComponent
Mini Compasses
Mini compass, dia: 16 mm
Catalog # 470003-496
 :  C1SG16-PK/12
Mini Compasses
Catalog # 470003-496
Mini Compasses
Mini Compasses
Mini compass, dia: 16 mm
Price
$34.60
Log in for availability
Pack of 12
Product Details & Documents
These small 16 mm diameter compasses are perfect for mapping out magnetic fields lines around a compass.
  • Highly accurate
  • Features a floating needle
These small compasses are ideal for mapping the magnetic field around a bar magnet.
Specifications
  • Description:
    Mini compass, dia: 16 mm