Microscope Slide Staining Dish with Rack
Microscope Slide Staining Dish with Rack, 20 Pcs., 41/8 x 33/8 x 23/4"
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 900200
Jump to:
Product Details & Documents
Dries Slides Efficiently with Room for Multiple Slides
- Quality slide making supplies
- Slide staining tools
This quick-draining glass rack holds ten single slides, 19 slides arranged in a zigzag pattern, or 20 slides back to back. It comes with a glass cover.
Documents
Specifications
- Description:Microscope Slide Staining Dish with Rack, 20 Pcs.
- Dimensions:41/8 x 33/8 x 23/4"